4-Piperidineethanol ≥96.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(2-Hydroxyethyl)piperidine, 2-(Piperidin-4-yl)ethanol
P0992-25G
P0992-5G
TCIAP0992-25GEA
232
EUR
TCIAP0992-25G
TCIAP0992-5G
4-Piperidineethanol ≥96.0% (by GC, titration analysis)
4-Piperidineethanol
Formula:
C₇H₁₅NO MW: 129.20 g/mol Pto. Ebullición: 228°C |
Número MDL:
MFCD00006008 Núm. CAS: 622-26-4 EINECS: 210-727-7 |
Resultados test especificaciones
Appearance | White or Colorless to Almost white or Almost colorless powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Nonaqueous Titration) | min. 96.0 % |
Melting point | 48.0 to 52.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...