Phosphorus pentoxide - methanesulphonic acid
Proveedor: TCI
|
Peligro
|
Sinónimos:
Eaton's Reagent
P1042-25G
P1042-500G
TCIAP1042-25GEA
16
EUR
TCIAP1042-25G
TCIAP1042-500G
Phosphorus pentoxide - methanesulphonic acid
Phosphorus pentoxide - methanesulphonic acid
MW:
141,95 g/mol |
Número MDL:
MFCD00070545 Núm. CAS: 39394-84-8 UN: 2922 ADR: 8,II |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Methane sulfonic acid (Neutralization Tit | 89.0 to 92.0 % |
Phosphorus pentoxide (Neutralization Tit. | 8.0 to 11.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...