2-Pyridylacetic acid hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl

P1076-25G P1076-5G
TCIAP1076-25GEA 132 EUR
TCIAP1076-25G TCIAP1076-5G
2-Pyridylacetic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
2-Pyridylacetic acid hydrochloride
Formula: C₇H₇NO₂
MW: 173,60 g/mol
Número MDL: MFCD00012812
Núm. CAS: 16179-97-8
EINECS: 240-316-8

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR