9-Phenanthreneboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI

P1093-1G P1093-5G
TCIAP1093-1GEA 16 EUR
TCIAP1093-1G TCIAP1093-5G
9-Phenanthreneboronic acid (contains varying amounts of Anhydride)
9-Phenanthreneboronic acid
Formula: C₁₄H₁₁BO₂
MW: 222.05 g/mol
Número MDL: MFCD00143524
Núm. CAS: 68572-87-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR