13-Fenil-1,4,7,10-tetraoxa-13-azaciclopentadecano ≥96.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 13-Phenyl-1,4,7,10-tetraoxa-13-azacyclopentadecane, N-Phenyl-13-aza-1,4,7,10-tetraoxacyclopentadecane, N-Phenylaza-15-crown 5-Ether

P1143-1G P1143-5G
TCIAP1143-1GEA 74 EUR
TCIAP1143-1G TCIAP1143-5G
13-Fenil-1,4,7,10-tetraoxa-13-azaciclopentadecano ≥96.0% (by GC, titration analysis)
13-Fenil-1,4,7,10-tetraoxa-13-azaciclopentadecano
Formula: C₁₆H₂₅NO₄
MW: 295.37 g/mol
Número MDL: MFCD00040513
Núm. CAS: 66750-10-5
EINECS: 266-470-6

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 44.0 to 47.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR