4-Pyridineethanol hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 4-(2-Hydroxyethyl)pyridine Hydrochloride, 2-(4-Pyridyl)ethanol Hydrochloride

P1293-25G P1293-5G P1293-1G
TCIAP1293-25GEA 860 EUR
TCIAP1293-25G TCIAP1293-5G TCIAP1293-1G
4-Pyridineethanol hydrochloride ≥98.0% (by HPLC, titration analysis)
4-Pyridineethanol hydrochloride
Formula: C₇H₉NO
MW: 159,61 g/mol
Número MDL: MFCD01321183
Núm. CAS: 383177-54-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 149.0 to 152.0 deg-C
Solubility in Water very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR