2-(Pyridin-4-yl)acetic acid hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Pyridylacetic acid hydrochloride

P1317-25G P1317-5G
TCIAP1317-25GEA 133 EUR
TCIAP1317-25G TCIAP1317-5G
2-(Pyridin-4-yl)acetic acid hydrochloride ≥98.0% (by HPLC, titration analysis)
2-(Pyridin-4-yl)acetic acid hydrochloride
Formula: C₇H₈ClNO₂
MW: 173,60 g/mol
Número MDL: MFCD00012827
Núm. CAS: 6622-91-9
EINECS: 229-576-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR