γ-Phenyl-γ-butyrolactone ≥97.0% (por GC)

Proveedor: TCI

Sinónimos: 5-Phenyltetrahydrofuran-2-one

P1321-25G P1321-5G
TCIAP1321-25GEA 358 EUR
TCIAP1321-25G TCIAP1321-5G
γ-Phenyl-γ-butyrolactone ≥97.0% (por GC)
gamma-Phenyl-gamma-butyrolactone
Formula: C₁₀H₁₀O₂
MW: 162,19 g/mol
Pto. Ebullición: 306°C
Número MDL: MFCD00005398
Núm. CAS: 1008-76-0
EINECS: 213-761-0

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 34.0 to 38.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR