Methyl (phenylthio)acetate

Proveedor: TCI
Atención

Sinónimos: (Phenylthio)acetic Acid Methyl Ester

P1334-25G P1334-5G
TCIAP1334-25GEA 145 EUR
TCIAP1334-25G TCIAP1334-5G
Methyl (phenylthio)acetate
Methyl (phenylthio)acetate
Formula: C₉H₁₀O₂S
MW: 182.24 g/mol
Pto. Ebullición: 102°C
Número MDL: MFCD00039805
Núm. CAS: 17277-58-6

Order Now

Resultados test especificaciones

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Specific gravity (20/20) 1.1710 to 1.1750
Refractive index n20/D 1.5580 to 1.5610


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR