Dimethyl-1,4-phenylenediacrylate ≥95.0% (por GC)
Proveedor: TCI
Sinónimos:
1,4-Phenylenediacrylic Acid Dimethyl Ester
P1338-5G
P1338-1G
TCIAP1338-5GEA
394
EUR
TCIAP1338-5G
TCIAP1338-1G
Dimethyl-1,4-phenylenediacrylate ≥95.0% (por GC)
Dimethyl-1,4-phenylenediacrylate
Formula:
C₁₄H₁₄O₄ MW: 246.26 g/mol |
Número MDL:
MFCD00185690 Núm. CAS: 7549-44-2 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...