Pyridostigmine bromide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
3-((Dimethylcarbamoyl)oxy)-1-methylpyridin-1-ium bromide, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide
P1339-25G
P1339-5G
TCIAP1339-25GEA
239
EUR
TCIAP1339-25G
TCIAP1339-5G
Pyridostigmine bromide ≥98.0% (by HPLC, titration analysis)
Pyridostigmine bromide
Formula:
C₉H₁₃BrN₂O₂ MW: 261,12 g/mol |
Número MDL:
MFCD00079283 Núm. CAS: 101-26-8 EINECS: 202-929-9 UN: 2811 ADR: 6.1,II Índice Merck: 14,07977 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...