5-Phenoxyvaleric acid 98,0 Titrimetric
Proveedor: TCI
|
Atención
|
P1357-5G
P1357-1G
TCIAP1357-5GEA
415
EUR
TCIAP1357-5G
TCIAP1357-1G
5-Phenoxyvaleric acid 98,0 Titrimetric
5-Phenoxyvaleric acid
Formula:
C₁₁H₁₄O₃ MW: 194.23 g/mol Pto. Ebullición: 165°C |
Número MDL:
MFCD02093487 Núm. CAS: 7170-40-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 63.0 to 66.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...