4-Pyridineethanethiol hydrochloride ≥97.0% (by titrimetric analysis)
Proveedor: TCI
Sinónimos:
4-(2-Mercaptoethyl)pyridine Hydrochloride, 2-(Pyridin-4-yl)ethanethiol hydrochloride, 2-(4-Pyridyl)ethanethiol Hydrochloride
P1373-25G
P1373-5G
TCIAP1373-25GEA
381
EUR
TCIAP1373-25G
TCIAP1373-5G
4-Pyridineethanethiol hydrochloride ≥97.0% (by titrimetric analysis)
4-Pyridineethanethiol hydrochloride
Formula:
C₇H₉NS MW: 175,67 g/mol |
Número MDL:
MFCD00956047 Núm. CAS: 6298-11-9 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (Iodometric Titration) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...