2-Pyridyl tribromomethyl sulphone ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI
Atención

Sinónimos: 2-((Tribromomethyl)sulphonyl)pyridine, 2-(Tribromomethanesulphonyl)pyridine

P1377-25G P1377-5G
TCIAP1377-25GEA 353 EUR
TCIAP1377-25G TCIAP1377-5G
2-Pyridyl tribromomethyl sulphone ≥98.0% (by HPLC, total nitrogen)
2-Pyridyl tribromomethyl sulphone
Formula: C₆H₄Br₃NO₂S
MW: 393,88 g/mol
Número MDL: MFCD02093493
Núm. CAS: 59626-33-4

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Solubility in Acetone almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR