Pyridoxine-3,4-dipalmitate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

P1395-25G P1395-5G
TCIAP1395-25GEA 146 EUR
TCIAP1395-25G TCIAP1395-5G
Pyridoxine-3,4-dipalmitate ≥98.0% (by HPLC, titration analysis)
Pyridoxine-3,4-dipalmitate
Formula: C₄₀H₇₁NO₅
MW: 646,01 g/mol
Número MDL: MFCD01632152
Núm. CAS: 992-42-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 88.0 to 92.0 deg-C
Solubility in hot THF almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR