L-Pipecolic acid ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
(S)-(-)-2-Piperidinecarboxylic Acid, H-HomoPro-OH, (S)-Piperidine-2-carboxylic acid, L-Pipecolinic Acid, L-Homoproline
P1404-1G
P1404-5G
TCIAP1404-1GEA
66
EUR
TCIAP1404-1G
TCIAP1404-5G
L-Pipecolic acid ≥98.0% (by titrimetric analysis)
L-Pipecolic acid
Formula:
C₆H₁₁NO₂ MW: 129,16 g/mol |
Número MDL:
MFCD00005981 Núm. CAS: 3105-95-1 EINECS: 221-462-1 Índice Merck: 14,07458 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -25.0 to -28.0 deg (C=4, H2O) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...