6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

P1436-1G P1436-5G
TCIAP1436-1GEA 65 EUR
TCIAP1436-1G TCIAP1436-5G
6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid ≥98.0% (by HPLC, titration analysis)
6-[2-Propyl-4-(4-pyridylazo)phenoxy]hexanoic acid
Formula: C₂₀H₂₅N₃O₃
MW: 355.44 g/mol
Núm. CAS: 312694-02-3

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 171.0 to 175.0 deg-C
Solubility in Acetic acid almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR