(S)-Propylene carbonate ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
P1486-25G
P1486-5G
TCIAP1486-25GEA
147
EUR
TCIAP1486-25G
TCIAP1486-5G
(S)-Propylene carbonate ≥98.0% (por GC)
(S)-Propylene carbonate
Formula:
C₄H₆O₃ Pto. Ebullición: 105°C |
Número MDL:
MFCD00798264 Núm. CAS: 51260-39-0 |
Resultados test especificaciones
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Specific rotation [a]20/D | -2.0 to -3.0 deg (neat) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...