Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulphonate ≥97.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Phenyl[2-(trimethylsilyl)phenyl]iodonium Triflate

P1620-1G P1620-5G
TCIAP1620-1GEA 150 EUR
TCIAP1620-1G TCIAP1620-5G
Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulphonate ≥97.0% (by HPLC, titration analysis)
Phenyl[2-(trimethylsilyl)phenyl]iodonium trifluoromethanesulphonate
Formula: C₁₆H₁₈F₃IO₃SSi
MW: 502,36 g/mol
Núm. CAS: 164594-13-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 97.0 %
Melting point 130.0 to 134.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR