Potassium trifluoro(trifluoromethyl)borate ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
Potassium (trifluoromethyl)trifluoroborate
P1692-1G
P1692-25G
P1692-5G
TCIAP1692-1GEA
83
EUR
TCIAP1692-1G
TCIAP1692-25G
TCIAP1692-5G
Potassium trifluoro(trifluoromethyl)borate ≥98.0% (by titrimetric analysis)
Potassium trifluoro(trifluoromethyl)borate
Formula:
CHBF₆ MW: 175,91 g/mol |
Número MDL:
MFCD09038544 Núm. CAS: 42298-15-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (Ion exchange titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...