Paladio 5% on barium sulphate

Proveedor: TCI

P1702-25G P1702-5G
TCIAP1702-25GEA 741 EUR
TCIAP1702-25G TCIAP1702-5G
Paladio 5% on barium sulphate
Palladium
Formula: Pd
Número MDL: MFCD03613605
Núm. CAS: 7440-05-3
EINECS: 231-115-6
Índice Merck: 14,06989

Order Now

Resultados test especificaciones

Appearance Light gray to Brown to Black powder to crystal
Content (Palladium) 4.90 to 5.10 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR