D-Phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: (R)-Methyl 2-amino-3-phenylpropanoate hydrochloride, H-D-Phe-OMe.HCl, H-D-Phe-OMe·HCl

P1725-25G P1725-5G
TCIAP1725-25GEA 72 EUR
TCIAP1725-25G TCIAP1725-5G
D-Phenylalanine methyl ester hydrochloride ≥98.0% (by HPLC, titration analysis)
D-Phenylalanine methyl ester hydrochloride
Formula: C₁₀H₁₃NO₂
MW: 215,68 g/mol
Pto. de fusión: 160°C
Número MDL: MFCD00066112
Núm. CAS: 13033-84-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Argentmetric Titration) min. 98.0 %
Specific rotation [a]20/D -35.0 to -39.0 deg (C=2, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR