N-Phthaloyl-DL-glutamic anhydride ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Phthalimidoglutaric Anhydride
P1789-25G
P1789-5G
TCIAP1789-25GEA
459
EUR
TCIAP1789-25G
TCIAP1789-5G
N-Phthaloyl-DL-glutamic anhydride ≥98.0% (by titrimetric analysis)
N-Phthaloyl-DL-glutamic anhydride
Formula:
C₁₃H₉NO₅ MW: 259,22 g/mol |
Número MDL:
MFCD00005886 Núm. CAS: 3343-28-0 EINECS: 222-088-1 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Morpholine Method) | min. 98.0 % |
Melting point | 202.0 to 208.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...