2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥98.0% (by HPLC)
Proveedor: TCI
|
Peligro
|
Sinónimos:
1-[(1,4-Benzodioxan-2-yl)carbonyl]piperazine Hydrochloride, 2-(1-Piperazinylcarbonyl)-1,4-benzodioxane Hydrochloride
P1890-25G
P1890-5G
TCIAP1890-25GEA
85
EUR
TCIAP1890-25G
TCIAP1890-5G
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride ≥98.0% (by HPLC)
2-(1-Piperazinylcarbonyl)-1,4-benzodioxane hydrochloride
Formula:
C₁₃H₁₆N₂O₃ MW: 284,74 g/mol Pto. de fusión: 263°C |
Número MDL:
MFCD00729051 Núm. CAS: 70918-74-0 EINECS: 415-660-9 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...