4-Propoxycinnamic acid ≥97.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-(4-Propoxyphenyl)acrylic Acid
P1898-25G
P1898-5G
TCIAP1898-25GEA
429
EUR
TCIAP1898-25G
TCIAP1898-5G
4-Propoxycinnamic acid ≥97.0% (by GC, titration analysis)
4-Propoxycinnamic acid
Formula:
C₁₂H₁₄O₃ MW: 206.24 g/mol |
Número MDL:
MFCD00995149 Núm. CAS: 69033-81-4 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...