Polyethylene glycolbis(3-aminopropyl)ether
Proveedor: TCI
P1948-1G
P1948-5G
TCIAP1948-1GEA
28
EUR
TCIAP1948-1G
TCIAP1948-5G
Polyethylene glycolbis(3-aminopropyl)ether
Polyethylene glycolbis(3-aminopropyl)ether
Número MDL:
MFCD00197739 Núm. CAS: 34901-14-9 |
Resultados test especificaciones
Appearance | White to Light yellow to Dark green powder to lump |
Amine Value | 45.0 to 49.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...