3-(4-Pyridyl)acrylic acid ≥97.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 4-Pyridineacrylic Acid, 3-(Pyridin-4-yl)acrylic acid

P1972-1G P1972-5G
TCIAP1972-1GEA 55 EUR
TCIAP1972-1G TCIAP1972-5G
3-(4-Pyridyl)acrylic acid ≥97.0% (by GC, titration analysis)
3-(4-Pyridyl)acrylic acid
Formula: C₈H₇NO₂
MW: 149,15 g/mol
Número MDL: MFCD00040746
Núm. CAS: 5337-79-1
EINECS: 226-265-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR