Phentolamine mesylate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: Phentolamine Methanesulphonate, 3-(((4,5-Dihydro-1H-imidazol-2-yl)methyl)(p-tolyl)amino)phenol methanesulphonate, 2-[N-(m-Hydroxyphenyl)-p-toluidinomethyl]imidazoline Methanesulphonate, Phentolamine methanesulphonate salt

P1985-100MG P1985-1G
TCIAP1985-100MGEA 16 EUR
TCIAP1985-100MG TCIAP1985-1G
Phentolamine mesylate ≥98.0% (by HPLC, titration analysis)
Phentolamine mesylate
Formula: C₁₇H₁₉N₃O
MW: 377,46 g/mol
Número MDL: MFCD00134201
Núm. CAS: 65-28-1
EINECS: 200-604-6
Índice Merck: 14,07264

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 178.0 to 182.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR