1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
7-Chloro-4-[4-(3-[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propyl)-1-piperazinyl]quinoline tetraphosphate tetrahydrate, UNII-854D7K8LXB, Piperaquine tetraphosphate tetrahydrate
P1987-25G
P1987-5G
TCIAP1987-25GEA
86
EUR
TCIAP1987-25G
TCIAP1987-5G
1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate ≥98.0% (by HPLC, total nitrogen)
1,3-Bis[4-(7-chloro-4-quinolinyl)-1-piperazinyl]propane tetraphosphate tetrahydrate
Formula:
C₂₉H₃₂Cl₂N₆·4H₃PO₄.4H₂O MW: 999,552 g/mol Pto. de fusión: 252°C |
Número MDL:
MFCD11870901 Núm. CAS: 915967-82-7 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % (calcd.on anh.substance) |
Water | 5.8 to 8.6 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...