Fenantridin-6(5H)-ona ≥98.0% (by HPLC, total nitrogen)

Proveedor: TCI

P1998-1G P1998-5G
TCIAP1998-1GEA 22 EUR
TCIAP1998-1G TCIAP1998-5G
Fenantridin-6(5H)-ona ≥98.0% (by HPLC, total nitrogen)
Fenantridin-6(5H)-ona
Formula: C₁₃H₉NO
Número MDL: MFCD00004988
Núm. CAS: 1015-89-0
EINECS: 213-804-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0189e62b1daf648338a044ad8787fa3412c0c07&vid=d0189e62b1daf648338a044ad8787fa3412c0c07&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR