Probucol ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
4,4'-(Isopropylidendithio)bis[2,6-di-tert-butylphenol], 4,4'-(Propane-2,2-diylbis(sulphanediyl))bis(2,6-di-tert-butylphenol)
P2002-5G
P2002-25G
TCIAP2002-5GEA
39
EUR
TCIAP2002-5G
TCIAP2002-25G
Probucol ≥98.0% (by HPLC)
Probucol
Formula:
C₃₁H₄₈O₂S₂ MW: 516,84 g/mol |
Número MDL:
MFCD00079281 Núm. CAS: 23288-49-5 EINECS: 245-560-9 Índice Merck: 14,07755 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Melting point | 125.0 to 128.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...