3-Pyridinecarboxamide oxime ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Pyridylamidoxime, Nicotinamide Oxime
P2003-1G
P2003-5G
TCIAP2003-1GEA
33
EUR
TCIAP2003-1G
TCIAP2003-5G
3-Pyridinecarboxamide oxime ≥98.0% (by HPLC, titration analysis)
3-Pyridinecarboxamide oxime
Formula:
C₆H₇N₃O MW: 137,14 g/mol |
Número MDL:
MFCD00265955 Núm. CAS: 1594-58-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...