Promethazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
10-[2-(Dimethylamino)propyl]phenothiazine Hydrochloride, N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine hydrochloride
P2029-25G
P2029-100G
TCIAP2029-25GEA
52
EUR
TCIAP2029-25G
TCIAP2029-100G
Promethazine hydrochloride ≥98.0% (by HPLC, titration analysis)
Promethazine hydrochloride
Formula:
C₁₇H₂₀N₂S MW: 320,88 g/mol |
Número MDL:
MFCD00012652 Núm. CAS: 58-33-3 EINECS: 200-375-2 UN: 3077 ADR: 9,III Índice Merck: 14,07786 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...