Potassium trimethoxy(trifluoromethyl)borate ≥95.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
P2036-5G
P2036-25G
TCIAP2036-5GEA
86
EUR
TCIAP2036-5G
TCIAP2036-25G
Potassium trimethoxy(trifluoromethyl)borate ≥95.0% (by titrimetric analysis)
Potassium trimethoxy(trifluoromethyl)borate
Formula:
C₄H₉BF₃KO₃ Pto. de fusión: 118°C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD22124929 Núm. CAS: 626232-27-7 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (Ion exchange titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...