2,2-Dimethyl-N-pyridin-4-ylpropionamide ≥98.0% (por GC)
Proveedor: TCI
Sinónimos:
2,2-Dimethyl-N-pyridin-4-yl-propionamide, 2,2-Dimethyl-N-(4-pyridinyl)propanamide, N-(4-Pyridyl)pivalamide, 4-(2,2,2-Trimethylacetamido)pyridine, 2,2-Dimethyl-N-(4-pyridyl)propionamide
P2044-1G
P2044-5G
TCIAP2044-1GEA
57
EUR
TCIAP2044-1G
TCIAP2044-5G
2,2-Dimethyl-N-pyridin-4-ylpropionamide ≥98.0% (por GC)
2,2-Dimethyl-N-pyridin-4-ylpropionamide
Formula:
C₁₀H₁₄N₂O MW: 178,24 g/mol |
Número MDL:
MFCD00996248 Núm. CAS: 70298-89-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 131.0 to 135.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...