3-(1-Piperazinyl)-1,2-benzisothiazole ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

P2064-1G P2064-5G
TCIAP2064-1GEA 31 EUR
TCIAP2064-1G TCIAP2064-5G
3-(1-Piperazinyl)-1,2-benzisothiazole ≥98.0% (by GC, titration analysis)
3-(1-Piperazinyl)-1,2-benzisothiazole
Formula: C₁₁H₁₃N₃S
Temperatura de almacenaje: Nevera
Número MDL: MFCD04117970
Núm. CAS: 87691-87-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0a6990ec907b5dd856aff3f2660cfbf6db29d2a&vid=a0a6990ec907b5dd856aff3f2660cfbf6db29d2a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 89.0 to 93.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR