Potassium-p-tolyl sulphate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

P2091-1G P2091-5G
TCIAP2091-1GEA 93 EUR
TCIAP2091-1G TCIAP2091-5G
Potassium-p-tolyl sulphate ≥98.0% (by HPLC, titration analysis)
Potassium-p-tolyl sulphate
Formula: C₇H₈O₄S
Temperatura de almacenaje: Congelador
Número MDL: MFCD28100835
Núm. CAS: 91978-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33f3b7135fb8a739b3da8d1b1345251b7f084b5a&vid=33f3b7135fb8a739b3da8d1b1345251b7f084b5a&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR