4-(1-Pyrrolidinylcarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid
P2105-1G
P2105-5G
TCIAP2105-1GEA
47
EUR
TCIAP2105-1G
TCIAP2105-5G
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid
Formula:
C₁₁H₁₄BNO₃ MW: 219,05 g/mol |
Número MDL:
MFCD03411950 Núm. CAS: 389621-81-2 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...