Propylene glycol monostearate ≥83.0% (por GC)

Proveedor: TCI

P2163-25G P2163-500G
TCIAP2163-25GEA 17 EUR
TCIAP2163-25G TCIAP2163-500G
Propylene glycol monostearate ≥83.0% (por GC)
Propylene glycol monostearate
Formula: C₂₁H₄₂O₃
Número MDL: MFCD00021902
Núm. CAS: 1323-39-3
EINECS: 215-354-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 83.0 %
Melting point 41.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR