Nicotinamidine hydrochloride ≥98.0% (by HPLC)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Amidinopyridinium chloride, 3-Amidinopyridine hydrochloride, Pyridine-3-carboximidamide Monohydrochloride, Pyridine-3-carboximidamide hydrochloride, 3-Carbamimidoylpyridine Monohydrochloride
P2167-1G
P2167-5G
TCIAP2167-1GEA
19
EUR
TCIAP2167-1G
TCIAP2167-5G
Nicotinamidine hydrochloride ≥98.0% (by HPLC)
Nicotinamidine hydrochloride
Formula:
C₆H₈ClN₃ MW: 157,60 g/mol |
Número MDL:
MFCD00054326 Núm. CAS: 7356-60-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...