Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

P2182-5G P2182-25G
TCIAP2182-5GEA 63 EUR
TCIAP2182-5G TCIAP2182-25G
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate ≥98.0% (by GC, titration analysis)
Phenyl(4,6-dimethoxypyrimidin-2-yl)carbamate
Formula: C13H13N3O4
Número MDL: MFCD02947731
Núm. CAS: 89392-03-0
EINECS: 406-600-2
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=860ad6ce5cbc073e712625d3c243df7aaf4c88d5&vid=860ad6ce5cbc073e712625d3c243df7aaf4c88d5&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR