2-Phenylacetohydrazide 98,0 GC, T
Proveedor: TCI
|
Atención
|
Sinónimos:
Phenylacetic Hydrazide, Phenylacetic acid hydrazide
P2203-5G
P2203-25G
TCIAP2203-5GEA
46
EUR
TCIAP2203-5G
TCIAP2203-25G
2-Phenylacetohydrazide 98,0 GC, T
2-Phenylacetohydrazide
Formula:
C₈H₁₀N₂O MW: 150.181 g/mol |
Número MDL:
MFCD00007612 Núm. CAS: 937-39-3 |
|
Resultados test especificaciones
Appearance | White to Gray to Red powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Bromination Method) | min. 98.0 % |
Melting point | 115.0 to 119.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...