D-Pinitol ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 3-O-Methyl-D-chiro-inositol, (1R,2S,3R,4S,5S,6S)-6-Methoxycyclohexane-1,2,3,4,5-pentaol

P2219-100MG P2219-1G
TCIAP2219-100MGEA 16 EUR
TCIAP2219-100MG TCIAP2219-1G
D-Pinitol ≥98.0% (by HPLC)
D-Pinitol
Formula: C₇H₁₄O₆
MW: 194,18 g/mol
Pto. de fusión: 188°C
Número MDL: MFCD00216659
Núm. CAS: 10284-63-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=392a68587238d3e2648ef194e098b26d71974f61&vid=392a68587238d3e2648ef194e098b26d71974f61&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Specific rotation [a]20/D +62.0 to +66.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR