6-Phenylpicolinic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 6-Phenylpicolinic acid, 6-Phenylpyridine-2-carboxylic acid

P2320-1G P2320-5G
TCIAP2320-1GEA 113 EUR
TCIAP2320-1G TCIAP2320-5G
6-Phenylpicolinic acid ≥98.0% (by GC, titration analysis)
6-Phenylpicolinic acid
Formula: C₁₂H₉NO₂
MW: 199.2054 g/mol
Número MDL: MFCD04114206
Núm. CAS: 39774-28-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e03fd1feb5788b37e25444af3a909299bcf0d2c4&vid=e03fd1feb5788b37e25444af3a909299bcf0d2c4&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR