Ácido 1-fenilciclopropanocarboxilico ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
1-Phenyl-1-cyclopropanecarboxylic acid
P2324-5G
P2324-25G
TCIAP2324-5GEA
54
EUR
TCIAP2324-5G
TCIAP2324-25G
Ácido 1-fenilciclopropanocarboxilico ≥98.0% (by GC, titration analysis)
Ácido 1-fenilciclopropanocarboxilico
Formula:
C₁₀H₁₀O₂ |
Número MDL:
MFCD00001288 Núm. CAS: 6120-95-2 EINECS: 228-090-9 |
|
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 84.0 to 88.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...