5-Phenylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Phenyl-2-thiophenecarboxylic acid, 5-Phenyl-2-thenoic acid

P2347-250MG P2347-1G
TCIAP2347-250MGEA 41 EUR
TCIAP2347-250MG TCIAP2347-1G
5-Phenylthiophene-2-carboxylic acid ≥98.0% (by GC, titration analysis)
5-Phenylthiophene-2-carboxylic acid
Formula: C11H8O2S
MW: 204,2450 g/mol
Número MDL: MFCD01313432
Núm. CAS: 19163-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bbb1c38300ec7f15527d007bb18e6e2c3aef49f9&vid=bbb1c38300ec7f15527d007bb18e6e2c3aef49f9&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR