4-Phosphonobutyric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Peligro

Sinónimos: (3-Carboxypropyl)phosphonic acid

P2349-200MG P2349-1G
TCIAP2349-200MGEA 37 EUR
TCIAP2349-200MG TCIAP2349-1G
4-Phosphonobutyric acid ≥98.0% (by titrimetric analysis)
4-Phosphonobutyric acid
Formula: C₄H₉O₅P
Número MDL: MFCD00041414
Núm. CAS: 4378-43-2
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b7e91ad69732c03b6eba2687987bab1476988623&vid=b7e91ad69732c03b6eba2687987bab1476988623&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystaline
Purity (Neutralization titration) min. 98.0 %
Melting point 125.0 to 129.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR