4-(trans-4-Pentylcyclohexyl)phenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

P2363-5G P2363-1G
TCIAP2363-5GEA 50 EUR
TCIAP2363-5G TCIAP2363-1G
4-(trans-4-Pentylcyclohexyl)phenylboronic acid (contains varying amounts of Anhydride)
4-(trans-4-Pentylcyclohexyl)phenylboronic acid
Formula: C₁₇H₂₇BO₂
MW: 274,21 g/mol
Número MDL: MFCD09750938
Núm. CAS: 143651-26-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb787d52eda208dcc55f3fa451d1f41a22aba98b&vid=eb787d52eda208dcc55f3fa451d1f41a22aba98b&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 108.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR