4'-Propyl-4-biphenylboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: (4'-Propyl-[1,1'-biphenyl]-4-yl)boronic acid

P2387-1G P2387-5G
TCIAP2387-1GEA 55 EUR
TCIAP2387-1G TCIAP2387-5G
4'-Propyl-4-biphenylboronic acid (contains varying amounts of Anhydride)
4'-Propyl-4-biphenylboronic acid
Formula: C₁₅H₁₇BO₂
MW: 240,11 g/mol
Número MDL: MFCD01114670
Núm. CAS: 153035-56-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e7fa2caa5bac3e3297ac42713028fb529f227f2&vid=2e7fa2caa5bac3e3297ac42713028fb529f227f2&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 97.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR