4-(3-Pyridyl)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-(3-Pyridyl)benzeneboronic acid hydrochloride
P2392-1G
P2392-5G
TCIAP2392-1GEA
125
EUR
TCIAP2392-1G
TCIAP2392-5G
4-(3-Pyridyl)phenylboronic acid hydrochloride (contains varying amounts of Anhydride)
4-(3-Pyridyl)phenylboronic acid hydrochloride
Formula:
C₁₁H₁₀BNO₂·HCl MW: 235,47 g/mol |
Núm. CAS:
2377369-59-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...